Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL11 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550360

Catalog No. PA550360


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ARL11 protein.

This gene encodes a tumor suppressor related to the ADP-ribosylation factor (ARF) family of proteins. The encoded protein may play a role in apoptosis in a caspase-dependent manner. Polymorphisms in this gene have been associated with some familial cancers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARL11
Polyclonal
Unconjugated
ARL11
ADP ribosylation factor like GTPase 11; ADP-ribosylation factor like GTPase 11; ADP-ribosylation factor-like 11; ADP-ribosylation factor-like protein 11; ADP-ribosylation factor-like tumor suppressor protein 1; ARL11; ARLTS1; C730007L20Rik
Rabbit
Antigen affinity chromatography
RUO
115761
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q969Q4
ARL11
Full length fusion protein of Human ARL11.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.