Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559340 Supplier Invitrogen™ Supplier No. PA559340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLWDVGGQAP LRASWKDYLE GTDILVYVLD STDEARLPES AAELTEVLND PNMAGV Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

This gene encodes a tumor suppressor related to the ADP-ribosylation factor (ARF) family of proteins. The encoded protein may play a role in apoptosis in a caspase-dependent manner. Polymorphisms in this gene have been associated with some familial cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL11
Gene Accession No. Q969Q4
Gene Alias ADP ribosylation factor like GTPase 11; ADP-ribosylation factor like GTPase 11; ADP-ribosylation factor-like 11; ADP-ribosylation factor-like protein 11; ADP-ribosylation factor-like tumor suppressor protein 1; ARL11; ARLTS1; C730007L20Rik
Gene Symbols ARL11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL11. Recombinant protein control fragment (Product #RP-97981).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115761
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.