Learn More
Invitrogen™ ARL11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589103
Description
Immunogen sequence: MGSVNSRGHK AEAQVVMMGL DSAGKTTLLY KLKGHQLVET LPTVGFNVEP LKAPGHVSLT LWDVGGQAPL RASWKDYLEG TDILVYVLDS TDEARLPESA AELTEVLNDP NMAGVPFLVL ANKQEAPDAL PLLKIRNRLS LERFQDHCWE LRGCSALTGE GLPEALQSLW SLLKSRSCMC LQARAHGAER GDSKRS; Positive Samples: Rat liver, Rat spleen.
This gene encodes a tumor suppressor related to the ADP-ribosylation factor (ARF) family of proteins. The encoded protein may play a role in apoptosis in a caspase-dependent manner. Polymorphisms in this gene have been associated with some familial cancers.
Specifications
ARL11 | |
Polyclonal | |
Unconjugated | |
ARL11 | |
ADP ribosylation factor like GTPase 11; ADP-ribosylation factor like GTPase 11; ADP-ribosylation factor-like 11; ADP-ribosylation factor-like protein 11; ADP-ribosylation factor-like tumor suppressor protein 1; ARL11; ARLTS1; C730007L20Rik | |
Rabbit | |
Affinity chromatography | |
RUO | |
364396 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.99 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q5BK71 | |
ARL11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-196 of human ARL11 (NP_6124591). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.