Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL13A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557925
Description
Immunogen sequence: KLVKENKCPC RVEPCSAIRN LERRNHQPIV EGLRWLLAVI DTCQLPPTSS ISISKNNTGS GERCSSHSFS T Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 59%.
The function of this protein remains unknown.
Specifications
ARL13A | |
Polyclonal | |
Unconjugated | |
ARL13A | |
ADP ribosylation factor like GTPase 13A; ADP-ribosylation factor like GTPase 13A; ADP-ribosylation factor-like 13; ADP-ribosylation factor-like 13A; ADP-ribosylation factor-like protein 13A; ARL13; ARL13A; dJ341D10.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
392509 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5H913 | |
ARL13A | |
Recombinant protein corresponding to Human ARL13A. Recombinant protein control fragment (Product #RP-95464). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction