Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104310 Supplier Invitrogen™ Supplier No. PA5104310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARL13B.

This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintenance of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL13B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARL13B
Gene Accession No. Q3SXY8, Q640N2
Gene Alias A530097K21Rik; A930014M17Rik; ADP ribosylation factor like GTPase 13B; ADP-ribosylation factor like GTPase 13B; ADP-ribosylation factor-like 13B; ADP-ribosylation factor-like 2-like 1; ADP-ribosylation factor-like protein 13B; ADP-ribosylation factor-like protein 2-like 1; Arl13b; Arl2l1; ARL2-like protein 1; C530009C10Rik; hnn; JBTS8; Protein hennin
Gene Symbols ARL13B
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARL13B(Accession Q3SXY8), corresponding to amino acid residues R20-F70.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200894, 304037, 487934, 68146
Target Species Human, Mouse, Rat, Canine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.