Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566639
Description
Immunogen sequence: LHSALQHPQLC TLPFLILANH QDKPAARSVQ EIKKYFELEP LARGKRWILQ PCSLDDMDAL KDSFSQLINL LEEKDHEAVR M Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
ADP-ribosylation factors (ARFs) are highly conserved guanine nucleotide binding proteins that enhance the ADP-ribosyltransferase activity of cholera toxin. ARFs are important in eukaryotic vesicular trafficking pathways and they play an essential role in the activation of phospholipase D (PC-PLD). ARL15 (ADP-ribosylation factor-like 15), is a member of the ARF family of the Ras superfamily of small GTPases that are known to be involved in multiple regulatory pathways altered in human carcinogenesis. Other members of this protein family include ARL11, thought to be a tumor suppressor and may play a role in the regulation of apoptosis, ARL2 which plays an important role in microtubule dynamics and cell cycle progression, and ARL4, which may play a role in neurogenesis during embryonic development. The gene encoding ARL15 maps to chromosome 5p15 in humans. At least three isoforms of ARL15 are known to exist.
Specifications
ARL15 | |
Polyclonal | |
Unconjugated | |
Arl15 | |
A430036I03; ADP ribosylation factor like GTPase 15; ADP-ribosylation factor like GTPase 15; ADP-ribosylation factor related protein 2; ADP-ribosylation factor-like 15; ADP-ribosylation factor-like protein 15; ADP-ribosylation factor-related protein 2; ARF-related protein 2; ARFRP2; Arl15; C230032K13Rik; FLJ20051 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54622 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXU5 | |
Arl15 | |
Recombinant Human ARL15. Recombinant protein control fragment (Product #RP-105289). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction