Learn More
Invitrogen™ ARL17A Polyclonal Antibody

Description
The antibody detects endogenous levels of total ARL17A protein.
Specifications
Specifications
| Antigen | ARL17A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ARL17A |
| Gene Accession No. | Q8IVW1 |
| Gene Alias | ADP ribosylation factor like GTPase 17A; ADP-ribosylation factor 7; ADP-ribosylation factor 7 variant; ADP-ribosylation factor-like 17-like; ADP-ribosylation factor-like protein 17; ARF1P2; ARF1P2 ARL17A; ARL17A; ARL17B; ARL17P1; PRO2667 |
| Gene Symbols | ARL17A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.