Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL17A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561339 Supplier Thermo Scientific Supplier No. PA561339
Add to Cart
Add to Cart

ARL17A Polyclonal Antibody

ARL17A Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL17A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL17A
Gene Accession No. Q8IVW1
Gene Alias ADP ribosylation factor like GTPase 17A; ADP-ribosylation factor 7; ADP-ribosylation factor 7 variant; ADP-ribosylation factor-like 17-like; ADP-ribosylation factor-like protein 17; ARF1P2; ARF1P2 ARL17A; ARL17A; ARL17B; ARL17P1; PRO2667
Gene Symbols ARL17A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL17A. Recombinant protein control fragment (Product #RP-104408).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.