Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583729
Description
Immunogen sequence: DLPGALSSNA IREVLELDSI RSHHWCIQGC SAVTGENLLP GIDWLLDDIS SRIFTA.
This gene encodes a small GTP-binding protein of the RAS superfamily which functions as an ADP-ribosylation factor (ARF). The encoded protein is one of a functionally distinct group of ARF-like genes.
Specifications
ARL2 | |
Polyclonal | |
Unconjugated | |
ARL2 | |
2610009M23Rik; ADP ribosylation factor like GTPase 2; ADP-ribosylation factor like GTPase 2; ADP-ribosylation factor-like 2; ADP-ribosylation factor-like protein 2; ADP-ribosylation-like 2; AI115441; ARFL2; arf-like protein 2; Arl184; Arl2; AW553335 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
402 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P36404 | |
ARL2 | |
Recombinant protein corresponding to Human ARL2. Recombinant protein control fragment (Product #RP-97564). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction