Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597915
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597915 Supplier Invitrogen™ Supplier No. PA597915
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARL2 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a small GTP-binding protein of the RAS superfamily which functions as an ADP-ribosylation factor (ARF). The encoded protein is one of a functionally distinct group of ARF-like genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ARL2
Gene Accession No. P36404
Gene Alias 2610009M23Rik; ADP ribosylation factor like GTPase 2; ADP-ribosylation factor like GTPase 2; ADP-ribosylation factor-like 2; ADP-ribosylation factor-like protein 2; ADP-ribosylation-like 2; AI115441; ARFL2; arf-like protein 2; Arl184; Arl2; AW553335
Gene Symbols ARL2
Host Species Rabbit
Immunogen Recombinant Human ADP-ribosylation factor-like protein 2 protein (19-184AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.