Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL2BP Antibody (001), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290277B
Description
ARL2BP Monoclonal specifically detects ARL2BP in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ARL2BP | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
1 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ARL2BP (Q9Y2Y0-1; Met1-His163). | |
0.1 mL | |
Signal Transduction | |
23568 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction