Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL2BP Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298591MFV450
Description
ARL2BP Polyclonal antibody specifically detects ARL2BP in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
ARL2BP | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2 | |
Produced in rabbits immunized with E. coli-derived Human ARL2BP fragment. | |
0.1 mL | |
Signal Transduction | |
23568 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction