Learn More
ARL2BP Mouse anti-Human, Clone: OTI1A10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800297

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2.GTP with high affinity but does not interact with ARL2.GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2.GTP but lack of ARL2 GTPase-activating protein activity.Specifications
ARL2BP | |
Monoclonal | |
Unconjugated | |
ARL2BP | |
BART, BART1, RP66 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23568 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y2Y0 | |
ARL2BP | |
Full length human recombit protein of human ARL2BP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.