Learn More
Invitrogen™ ARL2BP Polyclonal Antibody

Description
The antibody detects endogenous levels of total ARL2BP protein.
Specifications
Specifications
| Antigen | ARL2BP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ARL2BP |
| Gene Accession No. | Q4V8C5, Q9D385, Q9Y2Y0 |
| Gene Alias | 1700010P10Rik; 1700027H16Rik; 6330544B05Rik; ADP ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor like 2 binding protein; ADP-ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor-like 2 binding protein; ADP-ribosylation factor-like protein 2-binding protein; ADP-ribosylation-like 2 binding protein; AI482273; AI849834; Arf-like 2 binding protein BART1; ARF-like 2-binding protein; ARL2-binding protein; Arl2bp; Bart; BART1; binder of ARF2 protein 1; binder of Arl Two; binder of Arl2; QtsA-10827; retinitis pigmentosa 66 (autosomal recessive); RP66; unnamed protein product |
| Gene Symbols | ARL2BP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.