Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL2BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560227
Description
Immunogen sequence: MDALEGESFA LSFSSASDAE FDAVVGYLED IIMDDEFQLL QRNFMDKYYL EFEDTEENKL IYTPIFNEYI SLVEKYIEEQ LLQRI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2. GTP with high affinity but does not interact with ARL2. GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2. GTP but lack of ARL2 GTPase-activating protein activity.
Specifications
ARL2BP | |
Polyclonal | |
Unconjugated | |
ARL2BP | |
1700010P10Rik; 1700027H16Rik; 6330544B05Rik; ADP ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor like 2 binding protein; ADP-ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor-like 2 binding protein; ADP-ribosylation factor-like protein 2-binding protein; ADP-ribosylation-like 2 binding protein; AI482273; AI849834; Arf-like 2 binding protein BART1; ARF-like 2-binding protein; ARL2-binding protein; Arl2bp; Bart; BART1; binder of ARF2 protein 1; binder of Arl Two; binder of Arl2; QtsA-10827; retinitis pigmentosa 66 (autosomal recessive); RP66; unnamed protein product | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23568 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2Y0 | |
ARL2BP | |
Recombinant protein corresponding to Human ARL2BP. Recombinant protein control fragment (Product #RP-100104). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction