Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL2BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599094 Supplier Invitrogen™ Supplier No. PA599094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARL2BP Polyclonal Antibody for Western Blot, IHC (P), ELISA

ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2. GTP with high affinity but does not interact with ARL2. GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2. GTP but lack of ARL2 GTPase-activating protein activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL2BP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ARL2BP
Gene Accession No. Q9Y2Y0
Gene Alias 1700010P10Rik; 1700027H16Rik; 6330544B05Rik; ADP ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor like 2 binding protein; ADP-ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor-like 2 binding protein; ADP-ribosylation factor-like protein 2-binding protein; ADP-ribosylation-like 2 binding protein; AI482273; AI849834; Arf-like 2 binding protein BART1; ARF-like 2-binding protein; ARL2-binding protein; Arl2bp; Bart; BART1; binder of ARF2 protein 1; binder of Arl Two; binder of Arl2; QtsA-10827; retinitis pigmentosa 66 (autosomal recessive); RP66; unnamed protein product
Gene Symbols ARL2BP
Host Species Rabbit
Immunogen Recombinant Human ADP-ribosylation factor-like protein 2-binding protein (1-163AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23568
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.