Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL2BP Recombinant Rabbit Monoclonal Antibody (001)
ARL2BP Recombinant Rabbit Monoclonal Antibody (1)
Supplier: Thermo Scientific MA529038
Description
ARL2BP Recombinant Monoclonal Antibody for ICC/IF, ELISA

Specifications
ARL2BP | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9Y2Y0 | |
ARL2BP | |
Recombinant Human ARL2BP/BART1 protein (Met1-His163). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry | |
001 | |
Unconjugated | |
ARL2BP | |
1700010P10Rik; 1700027H16Rik; 6330544B05Rik; ADP ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor like 2 binding protein; ADP-ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor-like 2 binding protein; ADP-ribosylation factor-like protein 2-binding protein; ADP-ribosylation-like 2 binding protein; AI482273; AI849834; Arf-like 2 binding protein BART1; ARF-like 2-binding protein; ARL2-binding protein; Arl2bp; Bart; BART1; binder of ARF2 protein 1; binder of Arl Two; binder of Arl2; QtsA-10827; retinitis pigmentosa 66 (autosomal recessive); RP66; unnamed protein product | |
Rabbit | |
Protein A | |
RUO | |
23568 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction