Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL4D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563851 Supplier Invitrogen™ Supplier No. PA563851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGNHLTEMAP TASSFLPHFQ ALHVVVIGLD SAGKT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking. Mutations in this gene have been associated with Bardet-Biedl syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL4D
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL4D
Gene Accession No. P49703
Gene Alias 1110036H21Rik; ADP ribosylation factor like GTPase 4D; ADP-ribosylation factor like GTPase 4D; ADP-ribosylation factor-like 4D; ADP-ribosylation factor-like 5; ADP-ribosylation factor-like 6; ADP-ribosylation factor-like protein 4D; ADP-ribosylation factor-like protein 4L; ADP-ribosylation factor-like protein 5; Arf4l; Arfl4; ARL4D; Arl5; ARL6; AW456149; neuroprotective protein 2
Gene Symbols ARL4D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL4D. Recombinant protein control fragment (Product #RP-101956).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 379
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.