Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL4D Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519005
Description
This antibody is predicted to react with bovine, canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking. Mutations in this gene have been associated with Bardet-Biedl syndrome.
Specifications
| ARL4D | |
| Polyclonal | |
| Unconjugated | |
| ARL4D | |
| 1110036H21Rik; ADP ribosylation factor like GTPase 4D; ADP-ribosylation factor like GTPase 4D; ADP-ribosylation factor-like 4D; ADP-ribosylation factor-like 5; ADP-ribosylation factor-like 6; ADP-ribosylation factor-like protein 4D; ADP-ribosylation factor-like protein 4L; ADP-ribosylation factor-like protein 5; Arf4l; Arfl4; ARL4D; Arl5; ARL6; AW456149; neuroprotective protein 2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 379 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P49703 | |
| ARL4D | |
| Synthetic peptide sequence (KQDQPGALSAAEVEKR) corresponding to the internal amino acids of ARL4D (aa 136-152). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction