Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555479
Description
Immunogen sequence: QESLRSSWNT YYTNTEFVIV VVDSTDRERI SVTREELYKM LAHEDLRKAG LLIFANKQDV KECMTVAEIS QFLKLTSIKD HQWHIQACCA LTGEGLCQGL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
The protein encoded by this gene belongs to the ARF family of GTP-binding proteins. With its distinctive nuclear/nucleolar localization and interaction with HP1alpha, the protein is developmentally regulated and may play a role(s) in nuclear dynamics and/or signaling cascades during embryonic development. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
Specifications
| ARL5A | |
| Polyclonal | |
| Unconjugated | |
| ARL5A | |
| 2410015N24Rik; 2810410P22Rik; AA408731; ADP ribosylation factor like GTPase 5A; ADP-ribosylation factor like GTPase 5A; ADP-ribosylation factor-like 5; ADP-ribosylation factor-like 5A; ADP-ribosylation factor-like protein 5; ADP-ribosylation factor-like protein 5A; ARFLP5; ARL5; Arl5a; AW610751; T25534 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26225 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y689 | |
| ARL5A | |
| Recombinant protein corresponding to Human ARL5A. Recombinant protein control fragment (Product #RP-89615). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction