Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL5B Antibody (OTI2D7), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272463PEATT594
Description
ARL5B Monoclonal specifically detects ARL5B in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
ARL5B | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
ARL5B | |
Full length recombinant protein of human ARL5B (NP_848930) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Flow Cytometry | |
OTI2D7 | |
Flow Cytometry | |
ADP-ribosylation factor-like 5B, ADP-ribosylation factor-like 8, ADP-ribosylation factor-like protein 5B, ADP-ribosylation factor-like protein 8, ADP-ribosylation-like factor 8, ARL8 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
221079.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction