Learn More
Invitrogen™ ARL5B Polyclonal Antibody

Description
Predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | ARL5B |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ARL5B |
| Gene Accession No. | Q96KC2 |
| Gene Alias | 4930587A11Rik; ADP ribosylation factor like GTPase 5B; ADP-ribosylation factor like GTPase 5B; ADP-ribosylation factor-like 5B; ADP-ribosylation factor-like 8; ADP-ribosylation factor-like protein 5B; ADP-ribosylation factor-like protein 8; ADP-ribosylation-like factor 8; ARL5B; Arl8; AW146430 |
| Gene Symbols | ARL5B |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.