Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL5B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727380 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7380 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7380 Supplier Proteintech Group Inc Supplier No. 116941AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARL8 belongs to a family of proteins that are structurally similar to ADP-ribosylation factors. ARLs and ARFs are part of the RAS superfamily of regulatory GTPases.

Specifications

Antigen ARL5B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ARL5B
Gene Accession No. Q96KC2, Q9D4P0
Gene Alias ARL5B, ARL8
Gene Symbols Arl5b
Host Species Rabbit
Immunogen ARL5B Fusion Protein Ag2309
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221079, 75869
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.