Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548281
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548281 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548281 Supplier Invitrogen™ Supplier No. PA548281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARL5C Polyclonal Antibody for Western Blot

Binds and exchanges GTP and GDP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL5C
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ARL5C
Gene Accession No. A6NH57, Q6P068
Gene Alias ADP ribosylation factor like GTPase 5C; ADP-ribosylation factor like GTPase 5C; ADP-ribosylation factor-like 12; ADP-ribosylation factor-like 5C; ADP-ribosylation factor-like 5C protein; ADP-ribosylation factor-like protein 12; ADP-ribosylation factor-like protein 5C; ARL12; ARL5C; BC065791; putative ADP-ribosylation factor-like protein 5C
Gene Symbols ARL5C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 34-61 amino acids from the N-terminal region of human ARL5C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217151, 390790
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.