Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564433 Supplier Invitrogen™ Supplier No. PA564433
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEESILPKTH FFMWDIVRPE AL Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 55%.

Binds and exchanges GTP and GDP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL5C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL5C
Gene Accession No. A6NH57
Gene Alias ADP ribosylation factor like GTPase 5C; ADP-ribosylation factor like GTPase 5C; ADP-ribosylation factor-like 12; ADP-ribosylation factor-like 5C; ADP-ribosylation factor-like 5C protein; ADP-ribosylation factor-like protein 12; ADP-ribosylation factor-like protein 5C; ARL12; ARL5C; BC065791; putative ADP-ribosylation factor-like protein 5C
Gene Symbols ARL5C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL5C. Recombinant protein control fragment (Product #RP-103681).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.