Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL6 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297182MFV500
Description
ARL6 Polyclonal antibody specifically detects ARL6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ARL6 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ADP-ribosylation factor-like 6, ADP-ribosylation factor-like protein 6, Bardet-Biedl syndrome 3 protein, BBS3, MGC32934, RP55 | |
Produced in rabbits immunized with E. coli-derived Human ARL6 fragment. | |
0.1 mL | |
Signal Transduction | |
84100 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction