Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100129 Supplier Invitrogen™ Supplier No. PA5100129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BBS3.

The protein encoded by this gene belongs to the ARF-like (ADP ribosylation factor-like) sub-family of the ARF family of GTP-binding proteins which are involved in regulation of intracellular traffic. Mutations in this gene are associated with Bardet-Biedl syndrome (BBS). Alternative splicing results in multiple transcript variants of this gene. A vision-specific transcript encoding a different protein has been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARL6
Gene Accession No. O88848, Q9H0F7
Gene Alias 1110018H24Rik; 2210411E14Rik; ADP ribosylation factor like GTPase 6; ADP-ribosylation factor-like 6; ADP-ribosylation factor-like protein 6; ADP-ribosylation-like 6; ADP-ribosylation-like factor homolog ARL6; Arl6; Bardet-Biedl syndrome 3 protein; BBS3; RP55
Gene Symbols ARL6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARL6(Accession Q9H0F7), corresponding to amino acid residues T50-L100.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363760, 56297, 84100
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.