Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL6IP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP214313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214313 0.1 mL
NB437960 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP214313 Supplier Novus Biologicals Supplier No. NBP214313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARL6IP4 Polyclonal specifically detects ARL6IP4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ARL6IP4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q66PJ3
Gene Alias ADP-ribosylation-like factor 6 interacting protein 4, Aip-4, ARL-6-interacting protein 4, HSP-975, HSVI binding protein, HSVI-binding protein, MGC814, SFRS20, splicing factor, arginine/serine-rich 20, splicing regulator SRrp38, SR-15, SR-25ADP-ribosylation factor-like protein 6-interacting protein 4, SRp25 nuclear protein, SRp25aip-4, SRrp37
Gene Symbols ARL6IP4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: KKGKEKAEAQQVEALPGPSLDQWHRSAGEEEDGPVLTDEQKSRIQAMKPMTKEEWDARQSIIRKVVDPETGRTRLIKGDGEVLEEIVTKERHREINKVGVA
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51329
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.