Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL6IP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562548 Supplier Invitrogen™ Supplier No. PA562548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKGKEKAEAQ QVEALPGPSL DQWHRSAGEE EDGPVLTDEQ KSRIQAMKPM TKEEWDARQS IIRKVVDPET GRTRLIKGDG EVLEEIVTKE RHREINKVGV A Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

ARL6IP4 is a protein coding gene involved in modulating alternative pre-mRNA splicing with either 5' distal site activation or preferential use of 3' proximal site. In case of infection by Herpes simplex virus (HSVI), may act as a splicing inhibitor of HSVI pre-mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL6IP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL6IP4
Gene Accession No. Q66PJ3
Gene Alias AA408210; AA408365; ADP ribosylation factor like GTPase 6 interacting protein 4; ADP-ribosylation factor GTPase 6 interacting protein 4; ADP-ribosylation factor like GTPase 6 interacting protein 4; ADP-ribosylation factor-like 6 interacting protein 4; ADP-ribosylation factor-like protein 6-interacting protein 4; ADP-ribosylation-like factor 6 interacting protein 4; aip-4; ARL-6-interacting protein 4; Arl6ip4; HSP-975; HSVI binding protein; HSVI-binding protein; SFRS20; Splicing factor SRrp37; splicing factor, arginine/serine-rich 20; splicing regulator SRrp38; SR-15; SR-25; SRp25; SRp25 nuclear protein; SRrp37
Gene Symbols ARL6IP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL6IP4. Recombinant protein control fragment (Product #RP-97873).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51329
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.