Learn More
ARL6IP4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304665AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ARL6IP4 is a protein coding gene involved in modulating alternative pre-mRNA splicing with either 5′ distal site activation or preferential use of 3′ proximal site. In case of infection by Herpes simplex virus (HSVI), may act as a splicing inhibitor of HSVI pre-mRNA.Specifications
ARL6IP4 | |
Polyclonal | |
Unconjugated | |
ARL6IP4 | |
ADP-ribosylation factor GTPase 6 interacting protein 4, ADP-ribosylation factor like GTPase 6 interacting protein 4, ADP-ribosylation factor-like 6 interacting protein 4, ADP-ribosylation-like factor 6 interacting protein 4, aip-4, ARL-6-interacting protein 4, HSP-975, HSVI binding protein, HSVI-binding protein, SFRS20, splicing factor SRrp37, splicing factor, arginine/serine-rich 20, splicing regulator SRrp38, SR-15, SR-25, SRp25, SRp25 nuclear protein, SRrp37 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51329 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q66PJ3 | |
ARL6IP4 | |
Between 1 and 50 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.