Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL6IP4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568337
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8337 Supplier Bethyl Laboratories, Inc Supplier No. A304666A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

ARL6IP4 is a protein coding gene involved in modulating alternative pre-mRNA splicing with either 5′ distal site activation or preferential use of 3′ proximal site. In case of infection by Herpes simplex virus (HSVI), may act as a splicing inhibitor of HSVI pre-mRNA.

Specifications

Antigen ARL6IP4
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ARL6IP4
Gene Accession No. Q66PJ3
Gene Alias ADP-ribosylation factor GTPase 6 interacting protein 4, ADP-ribosylation factor like GTPase 6 interacting protein 4, ADP-ribosylation factor-like 6 interacting protein 4, ADP-ribosylation-like factor 6 interacting protein 4, aip-4, ARL-6-interacting protein 4, HSP-975, HSVI binding protein, HSVI-binding protein, SFRS20, splicing factor SRrp37, splicing factor, arginine/serine-rich 20, splicing regulator SRrp38, SR-15, SR-25, SRp25, SRp25 nuclear protein, SRrp37
Gene Symbols ARL6IP4
Host Species Rabbit
Immunogen Between 310 and 360
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51329
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.