Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556011 Supplier Invitrogen™ Supplier No. PA556011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIIYVVDSVD VDRLEEAKTE LHKVTKFAEN QGTPLLVIAN KQDLPKSLPV AEIEKQLALH ELIPATTYHV QPACAIIGEG LTEGMDKLYE MILKRRKSLK QKKKR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 64%.

ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL4C
Gene Accession No. P56559
Gene Alias ADP ribosylation factor like GTPase 4C; ADP ribosylation factor-like protein 7; ADP-ribosylation factor like GTPase 4C; ADP-ribosylation factor-like 4C; ADP-ribosylation factor-like 7; ADP-ribosylation factor-like protein 4C; ADP-ribosylation factor-like protein 7; ADP-ribosylation factor-like protein LAK; ARL4C; ARL7; LAK
Gene Symbols ARL4C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL7. Recombinant protein control fragment (Product #RP-88809).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.