Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100390 Supplier Invitrogen™ Supplier No. PA5100390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARL4C.

ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARL4C
Gene Accession No. P56559
Gene Alias ADP ribosylation factor like GTPase 4C; ADP ribosylation factor-like protein 7; ADP-ribosylation factor like GTPase 4C; ADP-ribosylation factor-like 4C; ADP-ribosylation factor-like 7; ADP-ribosylation factor-like protein 4C; ADP-ribosylation factor-like protein 7; ADP-ribosylation factor-like protein LAK; ARL4C; ARL7; LAK
Gene Symbols ARL4C
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARL4C(Accession P56559), corresponding to amino acid residues E148-R192.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10123
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.