Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL8A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214314
Description
ARL8A Polyclonal specifically detects ARL8A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARL8A | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q96BM9 | |
ARL8A | |
This antibody was developed against Recombinant Protein corresponding to amino acids: EKIEASKNELHNLLDKPQLQGIPVLVLGNKRDLPGA | |
0.1 mL | |
Signal Transduction | |
127829 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ADP-ribosylation factor-like 10B, ADP-ribosylation factor-like 8A, ADP-ribosylation factor-like protein 10B, ADP-ribosylation factor-like protein 8A, ARL10B, FLJ45195, GIE2, Novel small G protein indispensable for equal chromosome segregation 2, small G protein indispensable for equal chromosome segregation 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction