Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559205 Supplier Invitrogen™ Supplier No. PA559205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLQYSGKTTF VNVIASGQFN EDMIPTVGFN MRK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Plays a role in lysosome motility. In neurons, mediates the anterograde axonal long-range transport of presynaptic lysosome-related vesicles required for presynaptic biogenesis and synaptic function. May play a role in chromosome segregation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL8A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Arl8a
Gene Accession No. Q96BM9
Gene Alias 1110033P22Rik; ADP ribosylation factor like GTPase 8A; ADP-ribosylation factor like GTPase 8A; ADP-ribosylation factor-like 10B; ADP-ribosylation factor-like 8A; ADP-ribosylation factor-like protein 10B; ADP-ribosylation factor-like protein 8A; Arl10b; Arl8a; GIE2; Novel small G protein indispensable for equal chromosome segregation 2; RGD1565940; small G protein indispensable for equal chromosome segregation 2
Gene Symbols Arl8a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL8A. Recombinant protein control fragment (Product #RP-98820).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.