Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL8A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501731356 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1356 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1356 Supplier Proteintech Group Inc Supplier No. 170601AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in lysosomes motility. Alternatively, may play a role in chromosome segregation.

Specifications

Antigen ARL8A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARL8A
Gene Accession No. Q8VEH3, Q96BM9
Gene Alias ARL10B, ARL8A, GIE2
Gene Symbols Arl8a
Host Species Rabbit
Immunogen ARL8A Fusion Protein Ag10788
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127829, 68724
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.