Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100124 Supplier Invitrogen™ Supplier No. PA5100124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARL8B.

ARL8B may play a role in lysosome motility. may play a role in chromosome segregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL8B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARL8B
Gene Accession No. Q9CQW2, Q9NVJ2
Gene Alias 2610313E07Rik; 3100002J04Rik; ADP ribosylation factor like GTPase 8B; ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Arl10c; Arl8b; Gie1; novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1; Unknown (protein for MGC:137658)
Gene Symbols ARL8B
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARL8B(Accession Q9NVJ2), corresponding to amino acid residues N109-C159.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55207, 67166
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.