Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570286
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%.
ARL8B may play a role in lysosome motility. may play a role in chromosome segregation.
Specifications
ARL8B | |
Polyclonal | |
Unconjugated | |
ARL8B | |
2610313E07Rik; 3100002J04Rik; ADP ribosylation factor like GTPase 8B; ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Arl10c; Arl8b; Gie1; novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1; Unknown (protein for MGC:137658) | |
Rabbit | |
Affinity chromatography | |
RUO | |
55207 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NVJ2 | |
ARL8B | |
synthetic peptide directed towards the middle region of human ARL8B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction