Learn More
Invitrogen™ ARL8B Polyclonal Antibody

Description
ARL8B Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | ARL8B |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | ARL8B |
| Gene Accession No. | Q9CQW2, Q9NVJ2 |
| Gene Alias | 2610313E07Rik; 3100002J04Rik; ADP ribosylation factor like GTPase 8B; ADP-ribosylation factor like GTPase 8B; ADP-ribosylation factor-like 10C; ADP-ribosylation factor-like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; Arl10c; Arl8b; Gie1; novel small G protein indispensable for equal chromosome segregation 1; small G protein indispensable for equal chromosome segregation 1; Unknown (protein for MGC:137658) |
| Gene Symbols | ARL8B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.