Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570590 Supplier Invitrogen™ Supplier No. PA570590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 92%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Yeast: 79%; Zebrafish: 77%.

ARL9 is a member of the small GTPase protein family with a high degree of similarity to ARF proteins of the RAS superfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARL9
Gene Accession No. 0
Gene Alias 1700110A09Rik; ADP ribosylation factor like GTPase 9; ADP-ribosylation factor like GTPase 9; ADP-ribosylation factor-like 9; ADP-ribosylation factor-like protein 9; ARL9; Unknown (protein for MGC:153259); zgc:153259
Gene Symbols ARL9
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse ARL9.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 384185
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.