Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific RPR Test Cards
SDP

Catalog No. p-2827415 Shop All Arlington Scientific Products
Click to view available options
Quantity:
100 Case
25 Pack
300 Case
50 Pack
Format:
10-well
30-well
This item is not returnable. View return policy

Catalog No. 22-415-126

Supplier: Arlington Scientific 90601050

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with Arlington Scientific RPR Test Kits

Specifications

Detectable Analytes Reagin (Syphilis)
Format 10-well
Type RPR Test Card
For Use With (Application) ASI RPR test kit
Includes 18mm circle cards, 10-well
Quantity 50 Pack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.