Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arlington Scientific RPR Test Kit

Click to view available options
Quantity:
100 Tests
500 Tests
5000 Test 10-well Card
5000 Test 30-well Card
Description
- Available with or without controls
- Carbon antigen suspension
- Easy to interpret
- Sensitivity: >98%
- Specificity: >99%
- Color-coded, screw cap dropper vials—no glass ampules
- Liquid controls
- Three levels: reactive, weak reactive, and nonreactive
- Results on CAP and AAB surveys
- All testing materials provided
- NCCLS procedure sheets available
- CPD, CPDA-1 and EDTA may be used
Specifications
Specifications
Description | Detects “reagin” present in Serum of syphilitic patients |
Detectable Analytes | Reagin (Syphilis) |
Type | RPR Test Card |
For Use With (Application) | Syphilis Testing |
Includes | RPR Carbon Antigen 3 levels of Serum control 18mm circle cards spreading pipets Antigen dispensing bottle and needle, thirty-well cards |
Quantity | 5000 Test 10-well Card |
Specificity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction