Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck ARM-D™ (Antibiotic Resistance Monitoring and Detection) Kit, ampC
SDP

Catalog No. 23285152 Shop All Streck Laboratories Products
Change view
Click to view available options
ARM-D™ (Antibiotic Resistance Monitoring and Detection) Kit, ampC, Sufficient for 100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-285-152 ARM-D™ (Antibiotic Resistance Monitoring and Detection) Kit, ampC, Sufficient for 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-285-152 Supplier Streck Supplier No. 250032

missing translation for 'validMainframeMsg'

Targets six plasmid-mediated ampC resistance genes and can differentiate plasmid-mediated ampC β-lactamase resistance from chromosomal resistance.

  • Can be used with most 4-channel real-time PCR instruments
  • Validated for use with following systems: Applied Biosystems 7500 Fast, Applied Biosystems QuantStudio™ 7Flex, Bio-Rad™ CFX96 Touch™, QIAGEN Rotor-Gene Q and Streck

Specifications

Content And Storage Two multiplexed reactions, each targeting three gene families plus an internal control. Each PCR Mix 1 and 2 includes 250μL of primers and probe combinations for each target, so that 100 x 25μL PCR reactions can be performed. In addition, for each M
Product Type ARM-D™ (Antibiotic Resistance Monitoring and Detection) Kit, ampC
For Use With (Application) Gene Expression
No. of Reactions 100 Reactions
Includes Two multiplexed reactions, each targeting three gene families plus an internal control. Each PCR Mix 1 and 2 includes 250μL of primers and probe combinations for each target, so that 100 x 25μL PCR reactions can be performed.

For Research Use Only. Not for Use in Diagnostic Procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.