Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck ARM-D Kit, ampC (RUO) and Master Mix
SDP

Catalog No. 23285051 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
100 Reaction Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-285-051 100 Reaction Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-285-051 Supplier Streck Supplier No. 250044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Streck ARM-D kit, ampC is a multiplex real-time PCR kit for the rapid detection of AmpC β-lactamase resistance genes in Gram-negative bacteria. This comprehensive kit allows you to detect over 200 ampC allelic variants in the clinically relevant resistance gene families, CMY02, DHA, ACC, EBC, FOX and MOX, and differentiate between plasmid-mediated and chromosomal AmpC β-lactamase resistance.

  • Detects over 200 allelic variants in the CMY02, DHA, ACC, EBC, FOX and MOX gene families.
  • Compatible with most 4-channel real-time PCR thermal cyclers.
  • Protocol can be completed in less than one hour.
  • Contains positive controls for each gene target.
  • Detects gene variants not covered by cartridge-based kits.
  • Identifies resistance mechanisms before susceptibility results are available.

Warranty and Services

12 Month Dating

Specifications

Concentration 2X (Supermix), 10X (PCR Mix)
Content And Storage Store at -20 °C long term, 4 °C for short term
Control Template Endogenous Internal Control (16SrRNA gene)
Detection Method Primer-probe
Form Liquid
Format Individual vials of qPCR Primers/Probe, Supermix and controls
PCR Method Multiplex qPCR
Polymerase Taq DNA Polymerase
Technique Real-time PCR
For Use With (Equipment) 4-channel RT- PCR Thermal Cyclers
Gene Symbol RRS1
Genotyping Target AmpC β-lactamase resistance genes
Includes Label or Dye Yes
Kit Type qPCR DNA Detection Kit
Label or Dye Unlabeled
No. of Tests 100 Test
Optimal Reaction Temperature 60°C
Primer qPCR Primers
Product Line ARM-D
Product Type ARM-D Kit
Quantity 100 Reaction Kit
Research Category Antibiotic Resistance, Infectious Disease
Sufficient For 100 Reactions
Target Loci CMY02, DHA, ACC, EBC, FOX, MOX, 16S rRNA
Target Organism Class Gram-negative Bacteria
For Use With (Application) Gene Expression
Compatibility RT- PCR (qPCR) System
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Reaction Format Master Mix
Reagent Type PCR Master Mix, qPCR Primers, qPCR Probes
Sample Type Bacterial Isolates, Wastewater Samples
Starting Material DNA
Volume 25 μL
Includes Positive Controls for each target gene. Internal control (16S r RNA gene) included. Multiplex PCR premixes for ease of use. Step by step data acquisition and analysis guides.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.