Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck ARM-D Kit, β-Lactamase (RUO) and Mix
SDP

Catalog No. 23285052 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
100 Reaction Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-285-052 100 Reaction Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-285-052 Supplier Streck Supplier No. 250045
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Streck ARM-D Kit, β-Lactamase is a multiplex real-time PCR kit for the rapid detection of β-lactam antibiotic resistance genes in Gram-negative bacteria. This comprehensive kit allows you to detect over 450 allelic variants in 9 clinically relevant carbapenemases, extended-spectrum beta-lactamases (ESBLs) and plasmid-mediated ampC gene families, KPC, NDM, OXA-48, IMP, VIM, DHA, CMY-2, CTX-M-14 and CTX-M-15.

  • Detects over 450 allelic variants in the KPC, NDM, OXA-48, IMP, VIM, DHA, CMY-2, CTX-M-14 and CTX-M-15 gene families.
  • Compatible with most 4-channel real-time PCR thermal cyclers.
  • Protocol can be completed in less than one hour.
  • Contains positive controls for each gene target.
  • Includes a premixed 2X master mix (DNA Polymerase, dNTPs, MgCl2 and reaction buffer).
  • Detects gene variants not covered by cartridge-based kits.

Warranty and Services

12 Month Dating

Specifications

Concentration 2X (Supermix), 10X (PCR Mix)
Content And Storage Store at -20 °C long term, 4 °C for short term
Control Template Endogenous Internal Control (16SrRNA gene)
Detection Method TaqMan
Format Individual vials of qPCR Primers/Probe, Supermix and controls
PCR Method Multiplex qPCR
Technique RT- PCR, Hydrolysis Probe Assay
For Use With (Equipment) Real-Time PCR (qPCR) System
Gene Symbol blaKPC, blaNDM, blaOXA-48, blaIMP, blaVIM, blaDHA, blaCMY, blaCTX-M
Genotyping Target β-lactam antibiotic resistance genes
Includes Label or Dye Yes
Internal Probe Modification FAM (5'), HEX (5'), TEX615 (5'), TYE665 (5')
Kit Type qPCR DNA Detection Kit
No. of Tests 100 Test
Optimal Reaction Temperature 72°C
Primer PCR Primers
Product Line ARM-D
Product Type ARM-D Kit
Quantity 100 Reaction Kit
Research Category Infectious Disease
Sufficient For 100 Reactions
Target KPC, NDM, OXA-48, IMP, VIM, DHA, CMY-2, CTX-M-14, CTX-M-15
Target Loci KPC, NDM, OXA-48, IMP, VIM, DHA, CMY-2, CTX-M-14, CTX-M-15
Target Organism Class Gram-negative Bacteria
Target Specificity 99.5%
For Use With (Application) Gene Expression
Compatibility RT- PCR (qPCR) System
Final Product Type PCR Amplification
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Reaction Format Master Mix
Reagent Type PCR Assay
Sample Type Extracted and Purified DNA from Isolated Gram-negative Bacterial Colonies
Starting Material Extracted and Purified DNA
Includes Positive Controls for each target gene. Internal control (16S r RNA gene) included. Multiplex PCR premixes for ease of use. Step by step data acquisition and analysis guides.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.