Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARMC1 Antibody (OTI1D10), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271989AF647
Description
ARMC1 Monoclonal specifically detects ARMC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ARMC1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ARMC1 | |
Full length recombinant protein of human ARMC1 (NP_060590) produced in HEK293T cells. | |
0.1 mL | |
55156.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1D10 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Arcp, armadillo repeat containing 1, armadillo repeat-containing protein 1, FLJ10511 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction