Learn More
Invitrogen™ ARMC1 Monoclonal Antibody (3F7-A6)

Description
Peptide Sequence: MNSSTSTMSE EPDALSVVNQ LRDLAADPLN RRAIVQDQGC LPGLILFMDH PNPPVVHSAL LALRYLAECR ANREKMKGEL GMMLSLQNVI QKTTTPGETK LLASEIYDIL QSSNMADGDS FNEMNSRRRK AQFFLGTTNK RAKTVVLHID GLDDTSRRNL CEEALLKIKG VISFTFQMAV QRCVVRIRSD LKAEALASAI ASTKVMKAQQ VVKSESGEEM LVPFQDTPVE VEQNTELPDY LPEDESPTKE QDKAVSRVGS HPEGGASWLS TAANFLSRSF YW.
Specifications
Specifications
| Antigen | ARMC1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3F7-A6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ARMC1 |
| Gene Accession No. | Q9NVT9 |
| Gene Alias | Arcp; armadillo repeat containing 1; armadillo repeat-containing protein 1; ARMC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.