Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                ARMC10 Antibody, Novus Biologicals™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
| Antigen | ARMC10 | 
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Host Species | Rabbit | 
Description
ARMC10 Polyclonal specifically detects ARMC10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ARMC10 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| armadillo repeat containing 10, armadillo repeat-containing protein 10, FLJ37850, MGC3195, specific splicing variant involved in hepatocarcinogenesis, Splicing variant involved in hepatocarcinogenesis protein, SVHPNAS112 | |
| ARMC10 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | 
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| Q8N2F6 | |
| 83787 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LTDGSYDDVLNAEQLQKLLYLLESTEDPVIIERALITLGNNAAFSVNQAIIRELGGIPIVANKINHSNQSIKEKALNALNNLSVNVEN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
             
                                                 
                                                 
                                                 
                                                 
                                                 
                                                