Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARMC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100392 Supplier Thermo Scientific Supplier No. PA5100392
Add to Cart
Add to Cart

ARMC10 Polyclonal Antibody

ARMC10 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Armc10
Gene Accession No. B1WBW4, Q8N2F6, Q9D0L7
Gene Alias 2810037c14rik; armadillo repeat containing 10; armadillo repeat-containing protein 10; ARMC10; PNAS112; PNAS-112; PSEC0198; RGD1305915; specific splicing variant involved in hepatocarcinogenesis; splicing variant involved in hepatocarcinogenesis protein; Svh; SVH protein
Gene Symbols Armc10
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARMC10(Accession Q8N2F6), corresponding to amino acid residues Q131-Q181.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296758, 67211, 83787
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.