Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552783 Supplier Invitrogen™ Supplier No. PA552783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKLLLNLSEN PAMTEGLLRA QVDSSFLSLY DSHVAKEILL RVLTLFQNIK NCLKIEGHLA VQPTFTEGSL FFLLHGEECA QKIRALVDHH DAEVKEKVVT IIPKI Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.

This gene encodes a protein that contains an armadillo repeat and transmembrane domain. The encoded protein decreases the transcriptional activity of the tumor suppressor protein p53 through direct interaction with the DNA-binding domain of p53, and may play a role in cell growth and survival. Upregulation of this gene may play a role in hepatocellular carcinoma. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Armc10
Gene Accession No. Q8N2F6
Gene Alias 2810037c14rik; armadillo repeat containing 10; armadillo repeat-containing protein 10; ARMC10; PNAS112; PNAS-112; PSEC0198; RGD1305915; specific splicing variant involved in hepatocarcinogenesis; splicing variant involved in hepatocarcinogenesis protein; Svh; SVH protein
Gene Symbols Armc10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARMC10. Recombinant protein control fragment (Product #RP-90462).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83787
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.